Anti-Linker Mouse Polyclonal Antibody

Supplier: Enzo Life Sciences
ACC115-6000
76013-832EA 135 USD
76013-832
Anti-Linker Mouse Polyclonal Antibody
Antibodies
Recommended Applications: IHC, ISH (in situ hybridization)

  • Ready to use

Type: Primary
Antigen: Linker
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR