Human CD86 (from CHO cells)
Supplier: AdipoGen Life Sciences
ANC-579-820
102513-374EA 617.54 USD
102513-374
Human CD86 (from CHO cells)
Proteins and Peptides
Human CD86 (B7-2) is a costimulating ligand for CD28 and CD152 (CTLA-4). CD86 is expressed on activated B cells and blood monocytes.