Human CD27 (from CHO cells)
Supplier: AdipoGen Life Sciences
ANC-543-020
102513-360EA 617.54 USD
102513-360
Human CD27 (from CHO cells)
Proteins and Peptides
Human CD27 is a lymphocyte specific member of the TNF receptor family and is found primarily on peripheral blood T cells and on a subpopulation of B cells and NK cells. The ligand for CD27 is CD70, which is a member of the TNF ligand superfamily.