BX-795 ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: BX795, N-(3-((5-Iodo-4-((3-(thiophene-2-carboxamido)propyl)amino)pyrimidin-2-yl)amino)phenyl)pyrrolidine-1-carboxamide

CHM189-0005 CHM189-0010
76003-212EA 92.65 USD
76003-212 76003-214
BX-795 ≥97% (by HPLC)
BX-795

Inhibitor of PDK1, TBK1, IKK-ε, IRF3


BX795 was initially developed as a PDK1 inhibitor (direct competitive inhibitor IC50 = 11nM for PDK1) and has been shown to be a potent and relatively specific inhibitor of TBK1 and IKK-ε. It blocks the phosphorylation, nuclear translocation, and transcriptional activity of interferon regulatory factor 3. BX795 also is a potent inhibitor of cell growth of multiple cancer cell lines with IC50 values ranging from submicromolar amounts (0.368 µM to greater than 450 µM).

Formula: C₂₃H₂₆IN₇O₂S
MW: 591.5 g/mol
MDL Number: MFCD12546134
CAS Number: 702675-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fca3c8f71275c3aa3aa2d1ad93861ef791b5846&vid=00baacecaf4d89996e611024439cf19c12e149fb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR