KU55933 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: KU-55933, 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one

CHM150-0005 CHM150-0010
76003-096EA 86.11 USD
76003-096 76003-098
KU55933 ≥99% (by HPLC)
KU55933

Potent and selective ATP-competitive inhibitor of ATM


KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer.

Formula: C₂₁H₁₇NO₃S₂
MW: 395.49 g/mol
MDL Number: MFCD08276985
CAS Number: 587871-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7465ad788b50e879dddc5e0a365ae0deb6dc701f&vid=6c867d0448bc6d444eff8deb624125f375a5eb95&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR