Simvastatin sodium salt ≥97% (by HPLC)
Supplier: Enzo Life Sciences
CHM142-0005 CHM142-0010
76003-068EA 171.13 USD
76003-068 76003-070
Simvastatin sodium salt ≥97% (by HPLC)
Simvastatin sodium salt
Competitive inhibitor of HMG-CoA reductase
Simvastatin is marketed under the trade name Zocor™ and is often prescribed in combination with ezetimibe (Zetia) to treat dyslipidemia. This drug combination is known commercially as Vytorin or Inegy. Simvastatin also suppresses TNF-induced NF-κB activation, which potentiates apoptosis in human myeloid leukemia cells and thus, may be useful in treating cancer.
Suppresses TNF-induced NF-κB activation.
CAS Number: 101314-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b08b8dd8f3812d43f0ff30d292ab2ed53989b1fa&vid=ae09b29e56ae92dff8e7dcfe4fb34f83e86923db&prid=web&clid=SAPDAM