Dorsmorphin dihydrochloride ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
6-(4-(2-(Piperidin-1-yl)ethoxy)phenyl)-3-(pyridin-4-yl)pyrazolo[1,5-a]pyrimidine dihydrochloride, Dorsomorphin 2HCl
CHM141-0005
CHM141-0010
76003-064EA
177.67
USD
76003-064
76003-066
Dorsmorphin dihydrochloride ≥99% (by HPLC)
Dorsmorphin dihydrochloride
Potent and selective AMPK inhibitor.
Potent and selective AMPK inhibitor. Dorsomorphin also inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK-2), BMPR-IA (ALK-3), and BMPR-IB (ALK-6).
Formula:
C₂₄H₂₇Cl₂N₅O MW: 472.41 g/mol |
MDL Number:
MFCD11112197 CAS Number: 1219168-18-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...