WYE-125132 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

CHM140-0001 CHM140-0005
76003-060EA 120.99 USD
76003-060 76003-062
WYE-125132 ≥99% (by HPLC)
WYE-125132

Highly potent, ATP-competitive, and specific mTOR kinase inhibitor


A highly potent, ATP-competitive, and specific mTOR kinase inhibitor. Shows an anti-proliferative activity against a variety of tumor cell lines. WYE-132 causes inhibition of protein synthesis and cell size, induction of apoptosis, and cell cycle progression.


Antiproliferative.

MDL Number: MFCD22420813
CAS Number: 1144068-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d71d71d22db9e4c128ac6f49b97afb57a9da6da2&vid=6ff24274d5ebf03f6ab5565409c20e5668c4346a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR