IC87114 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-((6-Amino-9H-purin-9-yl)methyl)-5-methyl-3-(o-tolyl)quinazolin-4(3H)-one, IC-87114

CHM138-0005 CHM138-0010
76003-052EA 144.97 USD
76003-052 76003-054
IC87114 ≥99% (by HPLC)
IC87114

Cell permeable, selective inhibitor of PI 3-Kδ.

Formula: C₂₂H₁₉N₇O
MW: 397.4 g/mol
MDL Number: MFCD09970845
CAS Number: 371242-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df4fd13b9ab5a9572c3c80f4630fd62012a7e038&vid=f8db61decf154e1741d9121b6b87763413056104&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR