WYE-23 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

CHM136-0001 CHM136-0005
76003-044EA 134.07 USD
76003-044 76003-046
WYE-23 ≥99% (by HPLC)
WYE-23

Cell-permeable, potent, selective, and ATP-competitive mTOR inhibitor.

MW: 520.6 g/mol
CAS Number: 1062169-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b5bc731d9af6a9f758292256e36ea3d4ffefe58&vid=0856d0d8c3f0066ad37e8d3ddbac0f34a171f94b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR