Akt inhibitor VIII ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Akti-1/2, 1,3-Dihydro-1-[1-[4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenylmethyl]-4-piperidinyl]-2H-benzimidazol-2-one

CHM125-0001 CHM125-0005
76003-008EA 142.79 USD
76003-008 76003-010
Akt inhibitor VIII ≥98% (by HPLC)
Akt inhibitor VIII

Potent inhibitor of Akt1, Akt2 and Akt3 activity.


A cell-permeable compound that potently, selectively and reversibly inhibits Akt1/Akt2/Akt3 activity.

Formula: C₃₄H₂₉N₇O
MW: 551.65 g/mol
MDL Number: MFCD09026909
CAS Number: 612847-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df51de0bbff4d62f9a5dd1d49e849b487a7c5646&vid=0e03502adc415b7a67d93e07f7072f975f8eabf6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR