GSK-3 inhibitor XIII ≥99% (by HPLC)
Supplier: Enzo Life Sciences
CHM124-0001 CHM124-0005
76003-004EA 117.72 USD
76003-004 76003-006
GSK-3 inhibitor XIII ≥99% (by HPLC)
GSK-3 inhibitor XIII
Potent inhibitor of GSK-3 ATP-binding site.
CAS Number: 404828-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=870f095cbcf6cd35145e536245534395f10ebf19&vid=db7c3376450f2c45c46394a2d481dc489d3e069f&prid=web&clid=SAPDAM