MMP-13 inhibitor ≥99% (by HPLC)

Supplier: Enzo Life Sciences

CHM123-0001 CHM123-0005
76003-000EA 152.6 USD
76003-000 76003-002
MMP-13 inhibitor ≥99% (by HPLC)
MMP-13 inhibitor

Potent MMP-13 inhibitor.

Formula: C₂₂H₂₀F₂N₄O₂
MDL Number: MFCD09878513
CAS Number: 544678-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b3cdcac8fa8d090e5ba7efdfece8330f215e3aa&vid=542d326656354a278e2f21c37fd4b6ca83a9d322&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR