Chk2 inhibitor ≥97% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
Chk1 inhibitor II, Chk2 inhibitor I, 5-(2-Amino-5-oxo-1,5-dihydroimidazol-4-ylidine)-3,4,5,10-2H-azepino[3,4-b]indol-1-one
CHM115-0001
CHM115-0500
76002-498EA
513.39
USD
76002-498
76002-974
Chk2 inhibitor ≥97% (by HPLC)
Chk2 inhibitor
Cell-permeable, ATP-competitive, potent inhibitor of Chk2.
Displays anti-proliferative and anti-inflammatory properties. Chk2 inhibitor has also been reported to suppress the growth of leukemic T cells.
Formula:
C₁₅H₁₃N₅O₂ MW: 295.3 g/mol |
CAS Number:
724708-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...