SB590885 dihydrochloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
CHM113-0005
CHM113-0010
76002-490EA
177.67
USD
76002-490
76002-492
SB590885 dihydrochloride ≥98% (by HPLC)
SB590885 dihydrochloride
Potent B-Raf inhibitor.
Selective for B-Raf against 46 other kinases. SB 590885 dihydrochloride decreases anchorage-independent growth of melanoma cell lines. Inhibits ERK phosphorylation and proliferation in tumor cells expressing B-RafV600E.
Formula:
C₂₇H₂₇N₅O₂·2HCl |
CAS Number:
405554-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...