Bisindolylmaleimide I hydrochloride ≥99% (by HPLC)
Supplier: Enzo Life Sciences
CHM108-0001
CHM108-0005
76002-966EA
70.85
USD
76002-966
76002-968
Bisindolylmaleimide I hydrochloride ≥99% (by HPLC)
Bisindolylmaleimide I hydrochloride
Anti-inflammatory agent and protein kinase C inhibitor.
Enhanced, water soluble form of Bisindolylmaleimide I.
Formula:
C₂₅H₂₄N₄O₂·HCl MW: 452.5·36.5 g/mol |
CAS Number:
176504-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...