AdipoRon ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: SC-396658, 2-(4-Benzoylphenoxy)-N-[1-(phenylmethyl)-4-piperidinyl]acetamide, 2-(4-Benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide

CHM101-0005 CHM101-0010
76002-474EA 99.19 USD
76002-474 76002-476
AdipoRon ≥98% (by HPLC)
AdipoRon

Novel and orally active adiponectin receptor agonist for AdipoR1 and AdipoR2.


AdipoR agonist that binds to both AdipoR1 and AdipoR2.

Formula: C₂₇H₂₈N₂O₃
MW: 428.52 g/mol
MDL Number: MFCD08271060
CAS Number: 924416-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1335d48e27e8232fce64a8a92f9043eb317ad1c&vid=ac90f322903e2e8252b2cb0d6aaf51a752370b10&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR