BML-284 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-WN105-0005
BML-WN105-0025
76002-414EA
170.04
USD
76002-414
76002-416
BML-284 ≥98% (by TLC)
BML-284
Wnt activator
Wnt agonist that activates Wnt signaling in cellular assays and a Xenopus model. Induces β-catenin and TCF-dependent transcriptional activity in a dose-dependent manner (EC50=0.7μM). Wnt activation occured in a GSK-3β independent manner.
Formula:
C₁₉H₁₈N₄O₃ MW: 350.4 g/mol |
MDL Number:
MFCD09908087 CAS Number: 853220-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...