PNU-74654 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N'-((5-Methylfuran-2-yl)methylene)-2-phenoxybenzohydrazide

BML-WN101-0005 BML-WN101-0025
76002-406EA 116.63 USD
76002-406 76002-408
PNU-74654 ≥98% (by HPLC)
PNU-74654

Catenin blocker


Anti-tumor agent that disrupts the canonical Wnt/β-catenin pathway via blockage of the protein-protein interaction between β-catenin and TCF.

Formula: C₁₉H₁₆N₂O₃
MW: 320.3419 g/mol
MDL Number: MFCD12912438
CAS Number: 113906-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6fc5751a6e8727311f847f619dc3316e960710c&vid=dbdfab9ce3899ab0723ea2528ffc0036351bcc43&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR