Niltubacin
Supplier: Enzo Life Sciences
BML-GR363-0100 BML-GR363-0500
76002-374EA 107.91 USD
76002-374 76002-376
Niltubacin
Niltubacin
HDAC inhibitor
Synthetic.
Negative control for Tubacin (tubulin acetylation inducer), a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6. Concentration of niltubacin in cell culture experiments typically ranges from 2-20µM.
Synthetic.
Negative control for Tubacin (tubulin acetylation inducer), a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6. Concentration of niltubacin in cell culture experiments typically ranges from 2-20µM.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db0b9eab71d11e3da3861925bb681c3aef1f3061&vid=f4fbc261087379ca1782a78aa7b62a619c63a350&prid=web&clid=SAPDAM