Histone Deacetylase Inhibitor VIII ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR361-0001 BML-GR361-0005
76002-128EA 129.71 USD
76002-128 76002-368
Histone Deacetylase Inhibitor VIII ≥98% (by TLC)
Histone Deacetylase Inhibitor VIII

HDAC6 inhibitor


Selective HDAC6 inhibitor (IC50=0.002nM). IC50 values for other HDACs are 271, 252, 0.42, 6851 and 90.7 nM for HDACs 1,2,3,8 and 10 respectively. Potent inhibitor of the proliferation of a variety of pancreatic cancer cell lines (IC50s in the 0.1-0.6µM range).

Formula: C₂₂H₃₀N₄O₆
MDL Number: MFCD17010286
CAS Number: 1045792-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45565e7a1092844a24072993dae8e6b5cb79f66b&vid=efd3c03c8d163c520b2f60cb93ad0fa0c5e6cf40&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR