GW 9508 ≥98% (by TLC)
Supplier: Enzo Life Sciences
|
|
Synonyms:
GPR40 Agonist, GW9508, 4-[[(3-Phenoxyphenyl)methyl]amino]benzene propanoic acid, GW-9508, 3-(4-((3-Phenoxybenzyl)amino)phenyl)propanoic acid, 3-(4-(((3-(Phenoxy)phenyl)methyl)amino)phenyl)propanoic acid, Free Fatty Acid Receptor 1 Agonist I, FFA1 Agonist I
High potency synthetic agonist at GPR40.
First high potency synthetic agonist at GPR40. Enhanced insulin secretion in INS-1E cells and activates ATP-sensitive potassium channels in rat pancreatic beta cells (40 μM). It is used as a tool for exploring the biology of GPR40 mediated signaling and physiology.
Soluble in DMSO (15 mg/mL).
Formula:
C₂₂H₂₁NO₃ MW: 347.414 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09753282 CAS Number: 885101-89-3 UN: 3077 ADR: 9,III |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...