BI-D1870 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-((3,5-Difluoro-4-hydroxyphenyl)amino)-8-isopentyl-5,7-dimethyl-7,8-dihydropteridin-6(5H)-one

BML-EI407-0001 BML-EI407-0005
76002-100EA 98.1 USD
76002-100 76002-102
BI-D1870 ≥98% (by HPLC)
BI-D1870

S6 kinase inhibitor


Cell permeable, ATP-competitive inhibitor of the four isoforms of ribosomal S6 kinase (p90 RSK) (IC50's at 100µM ATP: RSK1 = 31nM, RSK2 = 24nM, RSK3 = 18nM, RSK4 = 15nM). Does not significantly inhibit 10 other AGC kinases, and over 40 other protein kinases at 100X concentrations. Acts on the N-terminal AGC kinase domain. Completely prevents PMA-induced TSC2 phosphorylation.

Formula: C₁₉H₂₃F₂N₅O₂
MW: 391.423 g/mol
MDL Number: MFCD11223662
CAS Number: 501437-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95110fcf55aa57f3a1f5d1df37d5dfeaa697cff7&vid=a1285e6997e559877937f1f69dfdadf7ac5d9c43&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR