ZK-756326 ≥98% (by TLC), white solid

Supplier: Enzo Life Sciences

Synonyms: ZK 756326, 2-(2-(4-(3-Phenoxybenzyl)piperazin-1-yl)ethoxy)ethan-1-ol, 2-[2-[4-[(3-Phenoxyphenyl)methyl]-1-piperazinyl]ethoxy]ethanol dihydrochloride

BML-CX101-0010 BML-CX101-0050
76002-064EA 150.42 USD
76002-064 76002-066
ZK-756326 ≥98% (by TLC), white solid
ZK-756326

CCR8 activator.


Novel small molecule agonist at the CCR8 chemokine receptor (EC₅₀ = 254 nM) with no activity at CCR1, CCR2B, CCR4, CCR5, CXCR2, CXCR3 or CXCR4. Dose dependently increases intracellular calcium and stimulates extracellular acidification in cells expressing human CCR8. Murine CCR8-expressing cells underwent chemotaxis on exposure to this agonist. Inhibits HIV fusion of cells expressing CD4 and CCR8. Small molecule CCR8 agonists represent useful tools for exploring CCR8 biology especially in experiments where the use of the natural protein agonist is impractical.


Soluble in water (5 mg/mL) or DMSO (4 mg/mL).

Formula: C₂₁H₂₈N₂O₃·2HCl
MW: 429.38 g/mol
MDL Number: MFCD09038571
CAS Number: 874911-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1327055066ef7b6b7d3e91850d26ef5b4cd0e5b&vid=af77cffcb77b8e0340a1158053695890937d76f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR