Lipid A 1 mg/mL in water (from E.coli), Serotype R515 (Re) ≥99.9% serotype J5 (Rc), TLRgrade™ ready-to-use non pyrogenic, sterile

Supplier: Enzo Life Sciences
Danger

581-200-L001 581-200-L002
76002-218EA 372.78 USD
76002-218 76203-050
Lipid A 1 mg/mL in water (from E.coli), Serotype R515 (Re) ≥99.9% serotype J5 (Rc), TLRgrade™ ready-to-use non pyrogenic, sterile
Lipid A

TLR4 activator.


Prepared by mild acid hydrolysis from purified E. coli R515 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form.


Activates Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice.

Order Now

Specification Test Results

Concentration 1 mg/mL
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity ≥99.9% (regarding protein or DNA contaminants, KDO not detectable)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR