KN-93 phosphate ≥98% (by 1H-NMR)

Supplier: Enzo Life Sciences

Synonyms: (E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide phosphate, (E)-KN-93 phosphate

430-127-M001 430-127-M005
76002-016EA 147.15 USD
76002-016 76002-018
KN-93 phosphate ≥98% (by 1H-NMR)
KN-93 phosphate

CaM kinase II inhibitor


Selective inhibitor of rat brain Ca2+/calmodulin-dependent protein kinase II. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A (PKA) activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Formula: C₂₆H₂₉ClN₂O₄S·H₃PO₄
MW: 501.0·98.0 g/mol
MDL Number: MFCD03788201
CAS Number: 1188890-41-6 | 139298-40-1 (free base)
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe3910654844a84fce4760702e32989e2b1cfc46&vid=22608058265f8862f82ee3ce5a8304cc6eb21208&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR