D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98%

Supplier: Enzo Life Sciences

Synonyms: Ins(1,4,5)P3.6K, IP3

307-007-M001 307-007-M005
76001-988EA 173.31 USD
76001-988 76001-990
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98%
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt

Product of phosphatidylinositol phospholipid hydrolysis.


Causes release of calcium from intracellular stores.


Hygroscopic and soluble in water.

Formula: C₆H₁₂K₃O₁₅P₃
Storage Temperature: Freezer
CAS Number: 103476-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3258accd826c12dfeefce4b935fc3d4c5f6a968&vid=1cb2c93d73097b3b8644969b230e5294ddba3cae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR