Anti-CD45R Monoclonal Antibody [clone: RA3-3A1/6.1]

Supplier: AdipoGen Life Sciences
ANC-720-020
102513-418EA 294.92 USD
102513-418
Anti-CD45R Monoclonal Antibody [clone: RA3-3A1/6.1]
Antibodies
Human CD45 is expressed on all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane tyrosine phosphatase which can exist in at least nine different isoforms resulting from tissue-specific alternative RNA splicing of exons 4-7 of a single gene coding for the various N-terminal peptide segments. CD45 plays an essential role in lymphocyte activation.

Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-3A1/6.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Rat IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR