Anti-CD3 Monoclonal Antibody (FITC) [clone: 145-2C11]

Supplier: AdipoGen Life Sciences
ANC-703-040
102513-392EA 346.15 USD
102513-392
Anti-CD3 Monoclonal Antibody (FITC) [clone: 145-2C11]
Antibodies
CD3 molecules associate with T cell receptors and provide intracellular signaling mechanism.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: FITC
Epitope:
Host:
Isotype: Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR