Anti-CD104 Monoclonal Antibody [clone: UMA9]

Supplier: AdipoGen Life Sciences
ANC-325-020
102512-956EA 346.15 USD
102512-956
Anti-CD104 Monoclonal Antibody [clone: UMA9]
Antibodies
Human CD104 (beta 4 integrin) associates with the alpha 6 integrin chain and binds to laminin and epiligrin and likely has cell-cell adhesion functions. CD104 is involved in signal transduction.

Type: Primary
Antigen: CD104
Clonality: Monoclonal
Clone: UMA9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR