Anti-p53 Monoclonal Antibody [clone: Pab240]

Supplier: AdipoGen Life Sciences
ANC-227-020
102512-584EA 294.92 USD
102512-584
Anti-p53 Monoclonal Antibody [clone: Pab240]
Antibodies
Human p53 is a tumor suppressor protein which is mutated in many cancers. p53 expression has been linked to apoptosis caused by DNA-damaging agents and may have a role in cell-mediated cytotoxicity.

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: Pab240
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR