Anti-CD64 Monoclonal Antibody [clone: 10.1]

Supplier: AdipoGen Life Sciences
ANC-216-580
102512-534EA 1784.77 USD
102512-534
Anti-CD64 Monoclonal Antibody [clone: 10.1]
Antibodies
Human CD64 is a high affinity receptor for monomeric human IgG1 and IgG3 which is expressed on macrophages, monocytes and gamma-interferon induced neutrophils. CD64 plays an important role in clearance of immune complexes and in antibody dependent cytoxicity.

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation:
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Dog, Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR