Anti-CD30 Monoclonal Antibody (Biotin) [clone: AC10]

Supplier: AdipoGen Life Sciences
ANC-179-030
102512-212EA 346.15 USD
102512-212
Anti-CD30 Monoclonal Antibody (Biotin) [clone: AC10]
Antibodies
Human CD30 is a member of the TNF receptor family and is expressed on mitogen activated B and T cells.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: AC10
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2bkappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR