Anti-CD16 Monoclonal Antibody (R-PE) [clone: 3G8]

Supplier: AdipoGen Life Sciences
ANC-165-050
102512-070EA 443.08 USD
102512-070
Anti-CD16 Monoclonal Antibody (R-PE) [clone: 3G8]
Antibodies
Human CD16 is a low affinity receptor for complexed IgG and occurs in two distinct forms, as a glycosyl phosphatidyl inositol (GPI) anchored protein in granulocytes and as an integral membrane protein on NK cells and macrophages.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR