Anti-CD3 Monoclonal Antibody [clone: UCHT1]

Supplier: AdipoGen Life Sciences
ANC-144-020
102511-856EA 294.92 USD
102511-856
Anti-CD3 Monoclonal Antibody [clone: UCHT1]
Antibodies
The human CD3/T cell receptor (TcR) complex is made up of the CD3 proteins (gamma, delta, 2 epsilon) and teh zeta-chain in association with either alpha/beta or gamma/delta proteins of the TcR. The TcR recognizes antigens in association with MHC molecules after which protein chains of the CD3 complex mediate activation signals triggered by TcR antigen binding. CD3 is expressed on greater than 95% of circulating human peripheral T cells.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR