LB Broth, Teknova
Supplier: TEKNOVA INC
L9135
L9130
L9145
100218-844EA
134.89
USD
100218-844
100219-108
100218-046
100218-394
LB Broth, Teknova
Media for Microbiology
Dehydrated Media for Microbiology
For the growth of
- Use 25 grams per Liter to reconstitute
- Ambient shipping conditions
- Store at room temperature
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), and 1.0% NaCl (10.0 g/L).
coli and other enteric bacteria.