Brady® Chemical, Biohazard, and Hazardous Material Signs: Non-Potable Water, Brady Worldwide
Supplier: Brady Worldwide
84467 84468 84313 84314 69407 22359 22360 69408 40847 40846 22283 22282 40923 40924
10810-344EA 15.59 USD
10810-344 10810-346 10810-374 10810-376 10810-456 10810-458 10810-460 10810-592 10810-602 10810-604 10810-654 10810-656 10810-678 10810-680
Brady® Chemical, Biohazard, and Hazardous Material Signs: Non-Potable Water, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T. Hazard Signs are included here.
Certificaciones: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.