Lauryl Sulphate Broth, HiMedia
Supplier: HiMedia
GM080-2.5KG GM080-500G GM080-5KG
75803-698EA 560.76 USD
75803-698 75803-700 75803-702
Lauryl Sulphate Broth, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Lauryl sulphate broth is a granulated media used for detection and enumeration of coliform bacteria in water, wastewater, dairy products and other food samples.