HiVeg™ Dey-Engley Neutralizing Broth Base, HiMedia Laboratories
Supplier: HiMedia
MV187-500G MV187-2.5KG MV187-5KG
61000-738EA 237.76 USD
61000-738 89228-202 89228-204
HiVeg™ Dey-Engley Neutralizing Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Dey-Engley neutralizing HiVeg media are used in disinfectant testing where neutralization of the antiseptics and disinfectants is important for determining its bactericidal activity.