Lidocaine hydrochloride

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219391710 0219391725 0219391780 0219391783
IC0219391710EA 96.45 USD
IC0219391710 IC0219391725 IC0219391780 IC0219391783
Lidocaine hydrochloride
Lidocaine hydrochloride

Lidocaine is the first amino amide–type local anestheticis.
Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.


Lidocaine is used topically to relieve itching, burning and pain from skin inflammations, injected as a dental anesthetic or as a local anesthetic for minor surgery a common local anesthetic and antiarrhythmic drug.

Formula: C₁₄H₂₃ClN₂O
Melting Pt: 72 - 82 °C
Storage Temperature: Ambient
CAS Number: 73-78-9
EINECS: 200-803-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Physical Appearance White Powder
pH 3.0 - 6.5 (1% aq soln)
Loss on Drying <8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR