Acetone ≥99.5% ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219383204 0219383280 0219383290 0219383291
IC0219383204EA 265.64 USD
IC0219383204 IC0219383280 IC0219383290 IC0219383291
Acetone ≥99.5% ACS
Acetone

Acetone is a commonly used material in the manufacture of various chemicals like methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid (via the ketene process), iodoform, and bromoform. Other products where acetone is utilized in manufacture include rayon, photographic films, and paint and varnish removers. In chemical synthesis, acetone has been utilized in the synthesis and isolation of both organic and inorganic compounds and complexes. These include t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles, and trispyrazolylborate platinum compounds. Acetone is also used in HPLC analysis of organic and inorganic species, including gold nanoparticle clusters and plant oils. Acetone has been utilized in protein crystallization.

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa) (lit.)
Melting Pt: –95.4 °C (lit.)
Density: 0.7905 - 0.793 g/cm3(Lit.)
Flash Pt: –17 °C (1.4 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8cb67b81017084c8b3ba5c12436ffb5a3ee6e00&vid=ebc855c6f8266baf1e17bee6e686e41aa8ef089f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance Clear, Colorless Liquid
Auto Ignition 540 °C(lit.)
Vapor Density 2.0 (air = 1)(Lit.)
Vapor Pressure 184 mmHg (20 °C)
Refractive Index n20/D 1.359(lit.)
Color APHA ≤10
Water Content ≤0.5%
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR