Calcium hydrogen orthophosphate, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

0219380480 0219380490 0219380491
IC0219380480EA 34.4 USD
IC0219380480 IC0219380490 IC0219380491
Calcium hydrogen orthophosphate, white powder
Calcium hydrogen orthophosphate

Calcium Phosphate is used in animal feeds, as a mineral supplement, as an antacid, in the manufacturing of glass, in dental products and in fertilizers. It has been used as a dietary supplement, and as an antacid.


Precaución: Research Use Only (RUO).

Formula: CaHPO₄
MW: 136.06 g/mol
Melting Pt: 370 (dec., lit.)
Density: ∼2.3 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR