Anthralin ≥99%, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one

0219150283 0219150291
IC19150283EA 73.07 USD
IC19150283 IC19150291
Anthralin ≥99%, yellow powder
Anthralin

Dithranol (1, 8 - dihydroxy-9-anthrone) is also known as anthralin. It is a vital topical antipsoriatic drugs. Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes.


Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile. Dithranol may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles. Dithranol may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.


Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes. Dithranol is being studied as a potential anti-psoriasis medication.


Soluble in chlororform (20 mg/ml - clear, bright yellow solution) and in fixed oilds; slightly soluble in ethanol (95%) and in ether; practically insoluble in water.

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Melting Pt: 175 - 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00053409
CAS Number: 1143-38-0
EINECS: 214-538-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 175 - 182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR