Magnesium chloride hexahydrate 99-102%, white crystals ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Magnesium dichloride

02191421.5 0219142101 0219142105 02191421.1
IC191421.5EA 130.06 USD
IC191421.5 IC19142101 IC19142105 IC02191421.1
Magnesium chloride hexahydrate 99-102%, white crystals ACS
Magnesium chloride hexahydrate

Magnesium chloride hexahydrate (MgCl2·6H2O) is an inorganic salt that can be generated from the acid leach solution of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in solar cookers. The result of altering poly (vinyl alcohol)(PVA) films with MgCl2·6H2O has been investigated. The ability of MgCl2·6H2O to reduce the flammability of cotton fabrics has been studied.


Magnesium chloride hexahydrate has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones.

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Melting Pt: 118 °C
Density: 2.316 - 2.33 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eace78374aed9e1a0f46c870036e8c0181505516&vid=eace78374aed9e1a0f46c870036e8c0181505516&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance White Crystals
Heavy Metals <5 ppm (as Pb),<5 ppm (Fe)
Insoluble Matter <0.005%
Nitrate (NO3) <0.001%
Phosphate (PO4) <5 ppm
Sulphate (SO4) <0.002%
Ammonium (NH4) <0.002%
Barium (Ba) <0.005%
Calcium (Ca) <0.01%
Manganese (Mn) <5 ppm
Potassium (K) <0.005%
Sodium (Na) <0.005%
Strontium (Sr) <0.005%
Heavy Metals (as Pb) <5 ppm
Iron (Fe) <5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR