1-Cetylpyridinium chloride, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Cetylpyridinium chloride, 1-Hexadecylpyridinium chloride, 1-Hexadecylpyridin-1-ium chloride

0219017725 0219017780 0219017790
IC0219017725EA 84.23 USD
IC0219017725 IC0219017780 IC0219017790
1-Cetylpyridinium chloride, crystalline powder
1-Cetylpyridinium chloride

Cetylpyridinium chloride is used is in some types of mouthwashes, toothpastes, lozenges, throat sprays, breath sprays, and nasal sprays. It is an antiseptic that kills bacteria and other microorganisms. It has been shown to be effective in preventing dental plaque and reducing gingivitis.

Formula: C₂₁H₃₈ClN
Melting Pt: 78…86 °C
Flash Pt: >93.3 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011731
CAS Number: 123-03-5
EINECS: 204-593-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4337305754d9eb65d06c5980e1aa6e1d48cfb604&vid=d24ebbeef40342908995a823ca633859f5739db0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance White Crystalline Powder
pH 5.0 - 7.0 (2% aq soln)
Solubility Very soluble in water, alcohol and chloroform; Slightly soluble in benzene and ether.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR