Gellan gum, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02180106.1 02180106.5 0218010601 0218010605
IC02180106.1EA 125.79 USD
IC02180106.1 IC02180106.5 IC0218010601 IC0218010605
Gellan gum, Powder
Gellan gum

Gellan gum is a water-soluble polysaccharide produced by Pseudomonas elodea, a bacterium.


Gelrite, gellan gum, is recommended as a gelling agent for use in place of agar in microbiological media. Gellan gum is used in a wide variety of immobilization matricies. As a food additive, gellan gum is used as a thickener, emulsifier, and stabilizer.

Storage Temperature: Ambient
MDL Number: MFCD00131909
CAS Number: 71010-52-1
EINECS: 275-117-5

Order Now

Specification Test Results

Physical Appearance Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR