NanoXact Silica Nanospheres, Silica Surface, nanoComposix
Supplier: NANOCOMPOSIX, INC.
SISN50-25M SISN50-100M SISN80-25M SISN80-100M SISN100-25M SISN100-100M SISN120-25M SISN120-100M SISN200-25M SISN200-100M SISN20-25M SISN20-100M SISN100-25M SISN100-500M SISN120-100M SISN120-1L SISN120-25M SISN120-500M SISN200-100M SISN200-1L SISN200-25M SISN200-500M SISN20-100M SISN20-1L SISN20-25M SISN20-500M SISN300-100M SISN300-1L SISN300-25M SISN300-500M SISN500-100M SISN500-1L SISN1000-100M SISN1000-1L SISN1000-25M SISN1000-500M SISN100-100M SISN100-1L SISN500-25M SISN500-500M SISN50-100M SISN50-1L SISN50-25M SISN50-500M SISN60-100M SISN60-1L SISN60-25M SISN60-500M SISN80-100M SISN80-1L SISN80-25M SISN80-500M
103369-686EA 220 USD
103369-686 103369-690 103369-694 103369-698 103369-702 103369-706 103369-710 103369-714 103369-726 103369-730 103370-098 103370-102 77680-622 77680-624 77680-626 77680-628 77680-630 77680-632 77680-634 77680-636 77680-638 77680-640 77680-642 77680-644 77680-646 77680-648 77680-650 77680-652 77680-654 77680-656 77680-658 77680-660 77680-610 77680-612 77680-614 77680-616 77680-618 77680-620 77680-662 77680-664 77680-666 77680-668 77680-670 77680-672 77680-674 77680-676 77680-678 77680-680 77680-682 77680-684 77680-686 77680-688
NanoXact Silica Nanospheres, Silica Surface, nanoComposix
Beads Microcarriers
High quality silica nanoparticles, provided with full COA for great value. Silica (silanol) surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Silica surface offers robust and versatile surface for a wide variety of applications
Spherical silica nanoparticles are available in 20 to 400 nm sizes, in water
Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Silica nanoparticles’ large surface area can be used for Absorption Studies, Nanosphere Lithography, and Coating applications. The silanol surface can be modified using standard silane chemistry to introduce specific functional groups or to create hydrophobic or fluorophillic surface coatings.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Información de suministro: Ships at ambient temperatures, via FedEx Priority Overnight (domestic).
Precaución: Do not freeze. Store at 25°C, bath sonicate if needed. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.