NanoXact Silica-Shelled Silver Nanospheres, 1 mg/ml, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AGSH50-1M AGSH50-5M AGSH50-10M AGSH50-10M AGSH50-1M AGSH50-5M
103370-078EA 300 USD
103370-078 103370-080 103370-082 77679-862 77679-864 77679-868
NanoXact Silica-Shelled Silver Nanospheres, 1 mg/ml, nanoComposix
Beads Microcarriers
Silver nanoparticles shelled with silica offer a versatile surface for a wide variety of applications.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration gold (1 mg/mL gold concentration)
  • Silica shell offers robust and versatile surface for a wide variety of applications
Spherical silver nanoparticles, 1 mg/mL silver concentration
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis. to meet the highest quality standards.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Información de suministro: Ships via FedEx Priority Overnight (domestic). Cold packaging included. Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Precaución: Do not freeze. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.