NanoXact Gold Nanospheres, Dried Powder, PVP Excipient, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUPD10-1MG AUPD10-5MG AUPD10-10MG AUPD10-10MG AUPD10-1MG AUPD10-5MG
103369-538EA 155 USD
103369-538 103369-540 103369-542 103369-550 103369-552 103369-554 103369-556 103370-232 103370-234 77680-112 77680-114 77680-118
NanoXact Gold Nanospheres, Dried Powder, PVP Excipient, nanoComposix
Beads Microcarriers
High quality gold nanoparticles. Dried powder, redispersible in polar solvents (water, alcohols).
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Dried powder dispersible in a wide variety of polar and aqueous solvents
Spherical gold nanoparticles, dried with PVP excipient to ensure particle stability so the materials redisperse without aggregation
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM images, TEM sizing, and UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Información de suministro: Ships at ambient temperatures, via FedEx Priority Overnight (domestic).
Precaución: Do not freeze. Store at 2 to 25 °C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.