Water, UltraPure, sterile filtered

Supplier: Rockland Immunochemical

Synonyms: Hydrogen oxide

MB-009-1000 MB-009-4000
RLMB-009-1000EA 82.54 USD
RLMB-009-1000 RLMB-009-4000
Water, UltraPure, sterile filtered
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR